DL-3-(BOC-Amino)-1-butanol 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-hydroxybutan-2-yl)carbamate

A112810-100MG A112810-1G A112810-250MG A112810-5G
76687-644EA 19 USD
76687-644 76687-646 76687-648 77570-704
DL-3-(BOC-Amino)-1-butanol 95%
DL-3-(BOC-Amino)-1-butanol

Precaución: for Research Use Only

Formula: C₉H₁₉NO₃
MDL Number: MFCD09031504
CAS Number: 146514-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR