5,6-Dichloro-1-ethyl-2-methylbenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloro-1-ethyl-2-methyl-1H-benzo[d]imidazole

A112077-1G A112077-5G
76687-636EA 41.38 USD
76687-636 76687-638
5,6-Dichloro-1-ethyl-2-methylbenzimidazole 98%
5,6-Dichloro-1-ethyl-2-methylbenzimidazole

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂N₂
MW: 229.10 g/mol
MDL Number: MFCD00022843
CAS Number: 3237-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR