3-(Dimethylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(N,N-Dimethylaminocarbonyl)phenylboronic acid, 3-(Dimethylcarbamoyl)benzeneboronic acid, (3-(Dimethylcarbamoyl)phenyl)boronic acid

A111739-10G A111739-1G A111739-250MG A111739-5G
76687-622EA 128.37 USD
76687-622 76687-624 76687-626 76687-628
3-(Dimethylcarbamoyl)phenylboronic acid 98%
3-(Dimethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BNO₃
MW: 193.01 g/mol
MDL Number: MFCD03412066
CAS Number: 373384-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR