1-(4-Chlorophenyl)-6,6-dimethyl-1,6-dihydro-1,3,5-triazine-2,4-diamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Cycloguanil HCl

A110956-1MG A110956-250MG A110956-100MG A110956-1G
76687-572EA 6 USD
76687-572 76687-574 77623-920 77878-840
1-(4-Chlorophenyl)-6,6-dimethyl-1,6-dihydro-1,3,5-triazine-2,4-diamine hydrochloride 97%
1-(4-Chlorophenyl)-6,6-dimethyl-1,6-dihydro-1,3,5-triazine-2,4-diamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅Cl₂N₅
MDL Number: MFCD00035308
CAS Number: 152-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR