2,3-Dimethyl-4-fluoronitrobenzene 98%

Supplier: Ambeed
Warning

A110781-100MG A110781-10G A110781-1G A110781-250MG A110781-25G A110781-5G
76687-544EA 5 USD
76687-544 76687-546 76687-548 76687-550 76687-552 76687-554
2,3-Dimethyl-4-fluoronitrobenzene 98%
2,3-Dimethyl-4-fluoronitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MDL Number: MFCD05663712
CAS Number: 1736-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR