5-Fluorouridine 97%

Supplier: Ambeed
Warning

A110675-10G A110675-1G A110675-5G A110675-25G A110675-100G
76687-536EA 96 USD
76687-536 76687-538 76687-540 76687-542 77623-916 77876-826
5-Fluorouridine 97%
5-Fluorouridine

Precaución: for Research Use Only

Formula: C₉H₁₁FN₂O₆
MDL Number: MFCD00036832
CAS Number: 316-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR