(E)-3-(2-((1S,4aS,5R,6R,8aS)-6-Hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylenedecahydronaphthalen-1-yl)ethylidene)furan-2(3H)-one 98%

Supplier: Ambeed
Warning

A110467-10MG A110467-1MG A110467-50MG
76687-516EA 7.43 USD
76687-516 76687-518 76687-520
(E)-3-(2-((1S,4aS,5R,6R,8aS)-6-Hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylenedecahydronaphthalen-1-yl)ethylidene)furan-2(3H)-one 98%
(E)-3-(2-((1S,4aS,5R,6R,8aS)-6-Hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylenedecahydronaphthalen-1-yl)ethylidene)furan-2(3H)-one

Precaución: for Research Use Only

Formula: C₂₀H₂₈O₄
MW: 332.43 g/mol
MDL Number: MFCD03093712
CAS Number: 134418-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR