Calcium-D gluconate 98%

Supplier: Ambeed
Warning

A110351-100G A110351-10G A110351-25G A110351-500G A110351-1000G
76687-502EA 9 USD
76687-502 76687-504 76687-506 76687-508 77876-823
Calcium-D gluconate 98%
Calcium-D gluconate

Precaución: for Research Use Only

Formula: HOH₂C(CH(OH))₄COOCa
MDL Number: MFCD00064209
CAS Number: 299-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR