Benzofuran-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A110181-250MG A110181-100MG A110181-1G
76687-492EA 80.63 USD
76687-492 77570-574 77623-912
Benzofuran-4-carboxylic acid 95%
Benzofuran-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₃
MW: 162.14 g/mol
MDL Number: MFCD10000600
CAS Number: 166599-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR