D-Galactal 95%

Supplier: Ambeed
Warning

A109730-25G A109730-5G A109730-1G
76687-464EA 284.33 USD
76687-464 77549-118 77599-578
D-Galactal 95%
D-Galactal

Precaución: for Research Use Only

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
MDL Number: MFCD00038067
CAS Number: 21193-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR