7-Bromo-6-chloroquinazolin-4(3H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 7-bromo-6-chloro-3,4-dihydroquinazolin-4-one

A109626-100G A109626-10G A109626-1G A109626-25G A109626-5G
76687-456EA 150 USD
76687-456 76687-458 76687-460 76687-462 77623-908
7-Bromo-6-chloroquinazolin-4(3H)-one 97%
7-Bromo-6-chloroquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂O
MDL Number: MFCD10000555
CAS Number: 17518-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR