1,2,3,4-Cyclobutanetetracarboxylic dianhydride 95%

Supplier: Ambeed
Warning

Synonyms: Cyclobuta[1,2-c:3,4-c']difuran-1,3,4,6(3aH,3bH,6aH,6bH)-tetraone

A109550-10G A109550-25G A109550-5G A109550-100G
76687-444EA 25 USD
76687-444 76687-446 76687-448 76687-450 77878-828
1,2,3,4-Cyclobutanetetracarboxylic dianhydride 95%
1,2,3,4-Cyclobutanetetracarboxylic dianhydride

Precaución: for Research Use Only

Formula: C₈H₄O₆
MDL Number: MFCD00004944
CAS Number: 4415-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR