N-(2-Bromobenzyl)-N,N-dimethylethanaminium 4-methylbenzenesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Bretylium tosylate

A109533-100MG A109533-10MG A109533-1MG A109533-250MG A109533-50MG A109533-5MG
76687-432EA 249.32 USD
76687-432 76687-434 76687-436 76687-438 76687-440 76687-442
N-(2-Bromobenzyl)-N,N-dimethylethanaminium 4-methylbenzenesulfonate 98%
N-(2-Bromobenzyl)-N,N-dimethylethanaminium 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₈H₂₄BrNO₃S
MW: 414.35 g/mol
MDL Number: MFCD00038731
CAS Number: 61-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR