(+)-3-Bromocamphor-8-sulfonic acid ammonium salt 97%

Supplier: Ambeed
Warning

Synonyms: Ammonium ((1R,3S,4S,7R)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulphonate

A109481-100G A109481-1G A109481-25G A109481-500G A109481-5G
76687-414EA 71 USD
76687-414 76687-416 76687-418 76687-420 76687-422
(+)-3-Bromocamphor-8-sulfonic acid ammonium salt 97%
(+)-3-Bromocamphor-8-sulfonic acid ammonium salt

Precaución: for Research Use Only

Formula: C₁₀H₁₈BrNO₄S
MW: 328.22 g/mol
MDL Number: MFCD00167438
CAS Number: 14575-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR