Arprinocid 95%

Supplier: Ambeed
Warning

Synonyms: Arpocox, 9-(2-Chloro-6-fluorobenzyl)-9H-purin-6-amine

A109289-100MG A109289-250MG A109289-1G A109289-5G A109289-25G
76687-410EA 5.31 USD
76687-410 76687-412 77570-534 77599-576 77623-902
Arprinocid 95%
Arprinocid

Precaución: for Research Use Only

Formula: C₁₂H₉ClFN₅
MW: 277.69 g/mol
MDL Number: MFCD00867210
CAS Number: 55779-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR