Boc-L-Tyr-ol 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Tyrosinol

A109066-1G A109066-5G
76687-406EA 8.49 USD
76687-406 76687-408
Boc-L-Tyr-ol 97%
Boc-L-Tyr-ol

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₄
MW: 267.33 g/mol
MDL Number: MFCD00270225
CAS Number: 220237-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR