Boc-Gly-N(OMe)Me 97%

Supplier: Ambeed
Warning

Synonyms: tert-butyl (2-(methoxy(methyl)amino)-2-oxoethyl)carbamate

A108829-100G A108829-10G A108829-1G A108829-25G A108829-500G A108829-5G
76687-386EA 204.76 USD
76687-386 76687-388 76687-390 76687-392 76687-394 76687-396
Boc-Gly-N(OMe)Me 97%
Boc-Gly-N(OMe)Me

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₄
MW: 218.3 g/mol
MDL Number: MFCD00674101
CAS Number: 121505-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR