5-Chloro-2-adamantanone 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloroadamantan-2-one

A108754-1G A108754-250MG A108754-25G A108754-5G
76687-370EA 23.34 USD
76687-370 76687-372 76687-374 76687-376
5-Chloro-2-adamantanone 98%
5-Chloro-2-adamantanone

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClO
MW: 184.66 g/mol
MDL Number: MFCD00798599
CAS Number: 20098-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR