3-Bromo-o-toluic acid 98%

Supplier: Ambeed
Warning

A108632-100G A108632-10G A108632-1G A108632-25G A108632-500G A108632-5G
76687-358EA 40 USD
76687-358 76687-360 76687-362 76687-364 76687-366 76687-368
3-Bromo-o-toluic acid 98%
3-Bromo-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MDL Number: MFCD00270097
CAS Number: 76006-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR