Butyl glycolate 97%

Supplier: Ambeed
Warning

A108539-100G A108539-10G A108539-1G A108539-5G A108539-25G A108539-500G
76687-350EA 37 USD
76687-350 76687-352 76687-354 76687-356 77570-494 77876-819
Butyl glycolate 97%
Butyl glycolate

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MDL Number: MFCD00042732
CAS Number: 7397-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR