Bismuth subsalicylate 98%

Supplier: Ambeed
Warning

Synonyms: Bismuth(III) subsalicylate, 2-Hydroxy-4H-benzo[d][1,3,2]dioxabismin-4-one

A107682-1000G A107682-100G A107682-25G A107682-500G
76687-292EA 128 USD
76687-292 76687-294 76687-296 76687-298
Bismuth subsalicylate 98%
Bismuth subsalicylate

Precaución: for Research Use Only

Formula: C₇H₅BiO₄
MDL Number: MFCD00085368
CAS Number: 14882-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR