1,1-Diethoxycyclohexane 98%

Supplier: Ambeed
Danger

Synonyms: Cyclohexanone diethyl ketal, Cyclohexanone diethyl acetal

A107156-5G A107156-25G
76687-284EA 36 USD
76687-284 77878-812
1,1-Diethoxycyclohexane 98%
1,1-Diethoxycyclohexane

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₂
MDL Number: MFCD00037723
CAS Number: 1670-47-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR