6-Nitroquinoline 98%

Supplier: Ambeed
Warning

A107018-100G A107018-10G A107018-1G A107018-25G A107018-500G A107018-5G
76687-272EA 122.01 USD
76687-272 76687-274 76687-276 76687-278 76687-280 76687-282
6-Nitroquinoline 98%
6-Nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₆N₂O₂
MDL Number: MFCD00006799
CAS Number: 613-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR