(R)-3-Aminoazepan-2-one 97%

Supplier: Ambeed
Warning

A106374-100MG A106374-1G A106374-250MG
76687-256EA 43.5 USD
76687-256 76687-258 76687-260
(R)-3-Aminoazepan-2-one 97%
(R)-3-Aminoazepan-2-one

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MW: 128.17 g/mol
MDL Number: MFCD00010201
CAS Number: 28957-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR