Boc-trans-3-hydroxy-L-proline 95%

Supplier: Ambeed
Warning

Synonyms: (2S,3S)-1-(tert-Butoxycarbonyl)-3-hydroxypyrrolidine-2-carboxylic acid, Boc-trans-3-OH-Pro-OH

A105472-100MG A105472-1G A105472-250MG A105472-5G
76687-230EA 28 USD
76687-230 76687-232 76687-234 77565-844
Boc-trans-3-hydroxy-L-proline 95%
Boc-trans-3-hydroxy-L-proline

Precaución: For research use only.

Formula: C₁₀H₁₇NO₅
MDL Number: MFCD04974546
CAS Number: 187039-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR