Boc-N-methyl-D-Valine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-N-Me-Val-OH, N-Boc-N-methyl-D-valine, (R)-2-((tert-Butoxycarbonyl)(methyl)amino)-3-methylbutanoic acid, Boc-N-Me-D-Val-OH

A104364-100G A104364-10G A104364-25G A104364-1G A104364-5G
76687-174EA 653 USD
76687-174 76687-176 76687-178 77570-266 77876-801
Boc-N-methyl-D-Valine 97%
Boc-N-methyl-D-Valine

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
MDL Number: MFCD00076996
CAS Number: 89536-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR