Cyclopentylacetic acid 98%

Supplier: Ambeed
Warning

A104161-10G A104161-1G A104161-25G A104161-5G
76687-166EA 17 USD
76687-166 76687-168 76687-170 76687-172
Cyclopentylacetic acid 98%
Cyclopentylacetic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MDL Number: MFCD00001387
CAS Number: 1123-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR