7-Bromo-4-chloroquinazoline 98%

Supplier: Ambeed
Warning

A103667-10G A103667-1G A103667-250MG A103667-25G A103667-5G
76687-144EA 209 USD
76687-144 76687-146 76687-148 76687-150 76687-152
7-Bromo-4-chloroquinazoline 98%
7-Bromo-4-chloroquinazoline

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂
MW: 243.4900 g/mol
MDL Number: MFCD06657216
CAS Number: 573675-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR