7-Chloro-3,4-dihydroquinolin-2(1H)-one 95%

Supplier: Ambeed
Warning

A103510-100MG A103510-1G A103510-250MG
76687-130EA 43.5 USD
76687-130 76687-132 76687-134
7-Chloro-3,4-dihydroquinolin-2(1H)-one 95%
7-Chloro-3,4-dihydroquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₈ClNO
MW: 181.62 g/mol
MDL Number: MFCD09033162
CAS Number: 14548-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR