1-Cyclopropylbutane-1,3-dione 97%

Supplier: Ambeed
Warning

A102990-1G A102990-5G
76687-104EA 8.49 USD
76687-104 77548-946
1-Cyclopropylbutane-1,3-dione 97%
1-Cyclopropylbutane-1,3-dione

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
MDL Number: MFCD00511094
CAS Number: 21573-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR