7-Nitroisatin 95%

Supplier: Ambeed
Warning

Synonyms: 7-Nitroindoline-2,3-dione

A102459-100G A102459-250MG A102459-5G
76687-088EA 361.77 USD
76687-088 76687-090 76687-092
7-Nitroisatin 95%
7-Nitroisatin

Precaución: for Research Use Only

Formula: C₈H₄N₂O₄
MW: 192.1300 g/mol
MDL Number: MFCD02066502
CAS Number: 112656-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR