cis-6-Nonen-1-ol 98%

Supplier: Ambeed
Warning

Synonyms: (Z)-6-Nonen-1-ol, (Z)-Non-6-en-1-ol

A102455-1G A102455-25G A102455-5G
76687-082EA 15.92 USD
76687-082 76687-084 76687-086
cis-6-Nonen-1-ol 98%
cis-6-Nonen-1-ol

Precaución: for Research Use Only

Formula: C₉H₁₈O
MW: 142.24 g/mol
MDL Number: MFCD00015388
CAS Number: 35854-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR