(+/-)-ɑ-Bisabolol 80%

Supplier: Ambeed
Danger

Synonyms: rel-(R)-6-Methyl-2-((R)-4-methylcyclohex-3-en-1-yl)hept-5-en-2-ol, 6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol

A102130-25G A102130-5G
76687-062EA 49.87 USD
76687-062 76687-064
(+/-)-ɑ-Bisabolol 80%
(+/-)-alpha-Bisabolol

Precaución: for Research Use Only

Formula: C₁₅H₂₆O
MW: 222.37 g/mol
MDL Number: MFCD00129082
CAS Number: 515-69-5
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR