4-Bromo-N-(2,5-diethoxyphenyl)benzenesulfonamide 97%

Supplier: Ambeed
Warning

A996199-1G A996199-250MG
76682-680EA 145.35 USD
76682-680 77623-446
4-Bromo-N-(2,5-diethoxyphenyl)benzenesulfonamide 97%
4-Bromo-N-(2,5-diethoxyphenyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₆H₁₈BrNO₄S
MW: 400.2874 g/mol
MDL Number: MFCD11729802
CAS Number: 1024505-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR