[4-(Benzyloxy)phenyl]methanamine hydrochloride 90%

Supplier: Ambeed
Warning

A996083-1G A996083-5G A996083-25G
76682-678EA 36 USD
76682-678 77784-737 77784-736
[4-(Benzyloxy)phenyl]methanamine hydrochloride 90%
[4-(Benzyloxy)phenyl]methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₆ClNO
MDL Number: MFCD16606045
CAS Number: 133100-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR