1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2,5-dihydro-1H-pyrrole-2,5-dione 95%

Supplier: Ambeed
Warning

A995918-100MG A995918-1G A995918-250MG A995918-5G
76682-658EA 46.68 USD
76682-658 76682-660 76682-662 76682-664
1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2,5-dihydro-1H-pyrrole-2,5-dione 95%
1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2,5-dihydro-1H-pyrrole-2,5-dione

Precaución: for Research Use Only

Formula: C₁₆H₁₈BNO₄
MW: 299.13 g/mol
MDL Number: MFCD20527119
CAS Number: 2096998-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR