1-Cyclopropyl-2-(triphenyl-lambda5-phosphanylidenE)ethan-1-one 98%

Supplier: Ambeed
Warning

A992923-1G A992923-25G A992923-5G
76682-632EA 13.8 USD
76682-632 76682-634 77623-422
1-Cyclopropyl-2-(triphenyl-lambda5-phosphanylidenE)ethan-1-one 98%
1-Cyclopropyl-2-(triphenyl-lambda5-phosphanylidenE)ethan-1-one

Precaución: for Research Use Only

Formula: C₂₃H₂₁OP
MW: 344.3860 g/mol
MDL Number: MFCD22124430
CAS Number: 7691-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR