Methyl 3-ethoxy-4-methoxybenzoate 95%

Supplier: Ambeed
Warning

A992452-100MG A992452-1G A992452-250MG A992452-5G
76682-624EA 39 USD
76682-624 76682-626 76682-628 76682-630
Methyl 3-ethoxy-4-methoxybenzoate 95%
Methyl 3-ethoxy-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MDL Number: MFCD06203845
CAS Number: 97966-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR