(1-(2-Methoxy-2-oxoethyl)-1H-pyrazol-4-yl)boronic acid 95%

Supplier: Ambeed
Warning

A992118-100MG A992118-1G A992118-250MG
76682-618EA 51 USD
76682-618 76682-620 76682-622
(1-(2-Methoxy-2-oxoethyl)-1H-pyrazol-4-yl)boronic acid 95%
(1-(2-Methoxy-2-oxoethyl)-1H-pyrazol-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₉BN₂O₄
MDL Number: MFCD28009361
CAS Number: 1146614-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR