2,5-Dihydroxy-4-methylbenzaldehyde 95%

Supplier: Ambeed
Warning

A991927-100MG A991927-1G A991927-25G
76682-612EA 18 USD
76682-612 76682-614 76682-616
2,5-Dihydroxy-4-methylbenzaldehyde 95%
2,5-Dihydroxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₃
MDL Number: MFCD24675617
CAS Number: 52010-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR