tert-Butyl ((3R,5S)-5-(trifluoromethyl)piperidin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A991887-100MG A991887-1G A991887-250MG A991887-5G
76682-604EA 113.52 USD
76682-604 76682-606 76682-608 76682-610
tert-Butyl ((3R,5S)-5-(trifluoromethyl)piperidin-3-yl)carbamate 95%
tert-Butyl ((3R,5S)-5-(trifluoromethyl)piperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₉F₃N₂O₂
MW: 268.2760 g/mol
MDL Number: MFCD23105832
CAS Number: 1523530-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR