9-([1,1'-Biphenyl]-3-yl)-3,6-dibromo-9H-carbazole 95%

Supplier: Ambeed
Warning

A990127-1G A990127-25G A990127-5G
76682-596EA 12.74 USD
76682-596 76682-598 77623-370
9-([1,1'-Biphenyl]-3-yl)-3,6-dibromo-9H-carbazole 95%
9-([1,1'-Biphenyl]-3-yl)-3,6-dibromo-9H-carbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₅Br₂N
MW: 477.1906 g/mol
MDL Number: MFCD30536711
CAS Number: 1221237-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR