2-Hydroxyethyl 4-methylbenzenesulfonate 95%

Supplier: Ambeed
Warning

Synonyms: Ethylene Glycol Monotosylate

A989220-1G A989220-5G
76682-590EA 233.4 USD
76682-590 77623-358
2-Hydroxyethyl 4-methylbenzenesulfonate 95%
2-Hydroxyethyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₉H₁₂O₄S
MW: 216.2542 g/mol
MDL Number: MFCD27950701
CAS Number: 42772-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR