2-(1-Carbamoylpiperidin-4-yl)acetic acid 95%

Supplier: Ambeed
Warning

A989140-100MG A989140-1G A989140-250MG
76682-584EA 83.82 USD
76682-584 76682-586 76682-588
2-(1-Carbamoylpiperidin-4-yl)acetic acid 95%
2-(1-Carbamoylpiperidin-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O₃
MW: 186.2084 g/mol
MDL Number: MFCD20638954
CAS Number: 279236-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR