5-Chloro-8-iodoh-imidazo[1,2-a]pyridine 98%

Supplier: Ambeed
Danger

Synonyms: 5-CHloro-8-iodoimidazo[1,2-a]pyridine

A986194-100MG A986194-1G A986194-250MG
76682-558EA 18 USD
76682-558 76682-560 76682-562
5-Chloro-8-iodoh-imidazo[1,2-a]pyridine 98%
5-Chloro-8-iodoh-imidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₄ClIN₂
MDL Number: MFCD14585310
CAS Number: 1031289-77-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR