2-Amino-6-methylphenol hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A985621-1G
A985621-250MG
A985621-5G
76682-552EA
50.93
USD
76682-552
76682-554
76682-556
2-Amino-6-methylphenol hydrochloride 98%
2-Amino-6-methylphenol hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₀ClNO MW: 159.6134 g/mol |
MDL Number:
MFCD21364897 CAS Number: 78886-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...