tert-Butyl N-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]carbamate 95%

Supplier: Ambeed
Warning

A984960-100MG A984960-1G A984960-250MG A984960-5G
76682-544EA 58.35 USD
76682-544 76682-546 76682-548 76682-550
tert-Butyl N-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]carbamate 95%
tert-Butyl N-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₈BNO₄
MW: 285.1874 g/mol
MDL Number: MFCD22383765
CAS Number: 1850305-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR