7,8-difluorodibenzo[b,e]thiepin-11(6H)-one 98%

Supplier: Ambeed
Warning

A838887-1G A838887-25G A838887-5G
76682-514EA 31.83 USD
76682-514 76682-516 77569-566
7,8-difluorodibenzo[b,e]thiepin-11(6H)-one 98%
7,8-difluorodibenzo[b,e]thiepin-11(6H)-one

Precaución: For research use only.

Formula: C₁₄H₈F₂OS
MW: 262.2745 g/mol
MDL Number: MFCD31706271
CAS Number: 2136287-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR