2-Bromo-N-isopropylbenzamide 98%

Supplier: Ambeed
Danger

A763794-1G A763794-25G A763794-5G
76682-498EA 22.28 USD
76682-498 76682-500 76682-502
2-Bromo-N-isopropylbenzamide 98%
2-Bromo-N-isopropylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO
MW: 242.1100 g/mol
MDL Number: MFCD00779656
CAS Number: 64141-90-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR