Actarit 99%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Acetamidophenyl)acetic acid, 4-Acetamidophenylacetic acid

A214764-1G A214764-250MG A214764-5G A214764-100G A214764-25G
76682-486EA 6.37 USD
76682-486 76682-488 77600-730 77622-262 77622-264
Actarit 99%
Actarit

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
MW: 193.20 g/mol
MDL Number: MFCD00196165
CAS Number: 18699-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR